Learn More
Brady™ Horizontal Safety Sign - No Access for Unauthorized Persons
Product Code. 16521330 Shop All Brady Products
Change view
Click to view available options
Dimensions (L x W):
150 x 450 mm
Unit Size:
Each
Product Code. Dimensions (L x W) Unit Size
16521330 150 x 450 mm Each
1 options
Product Code. 16521330 Supplier Brady™ Supplier No. 195925
  • ONLINE EXCLUSIVE PRICE
    226.00 SEK
    193.64 SEK / Paket med 1
    Save 32.36 SEK
    14% Av
Beräknad leverans: 10-12-2026
Safety sign for indicating mandatory action.
  • Improves workplace safety
Specifications
Alternate Part Number P/PIC209/FR570-PP-450x150mm/1-B
Material Polypropylene
Shape Rectangular
Languages French
Dimensions (L x W) 150 x 450 mm
Displayed Text Accès interdit aux personnes non autorisées
Color Red on White