Läs mer
Honeywell™ Howard Leight™ Viking™ Ear Muffs
Noise Reduction Rating (NRR):
29 dB
Stil:
Viking 1 Earmuffs
Förpackningsstorlek:
Varje
Beskrivning
- Fits with other PPE, including hard hats, visors, face shields, respirators and welding shields
- Dielectric construction won't deform
- Snap-in ear cushions
Specifikationer
Specifikationer
| Certifikat och efterlevnad | ANSI S.3.19-1974 |
| För användning med (applikation) | Agriculture and farming, building construction, forestry, general contracting, heavy construction, landscaping, lumber or wood products, metal fabrication Petrochemical Steel Mining Transportation Utility/Waste Management |
| Noise Reduction Rating (NRR) | 29 dB |
| Produkttyp | Ear Muffs |
| Dielektrisk | Yes |
| Stil | Viking 1 Earmuffs |
| Inkluderar | Inner ventilation |