missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXA7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567324
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: VNALFSKYTAG TSLFQNAEPT SCSFAPNSQR SGYGA Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.
Hox genes play a fundamental role in the development of the vertebrate central nervous system, heart, axial skeleton, limbs, gut, urogenital tract and external genitalia. HoxA2 protein may contribute to dorsal-ventral patterning and/or to the specification of neuronal phenotype. HoxA7 functions as a potent transcriptional repressor and its action as such requires several domains, including both activator and repressor regions. HoxA7 is expressed in the fetal liver, lung, skeletal muscle, kidney, pancreas and placenta.Specifications
HOXA7 | |
Polyclonal | |
Unconjugated | |
HOXA7 | |
ANTP; AV118143; homeo box A7; homeobox A7; homeobox A9; Homeobox gene A7; homeobox protein Hox 1.1; Homeobox protein Hox-1.1; homeobox protein HOX-1A; Homeobox protein Hox-A7; HOMEOBOX PROTEIN HOX-A7 (HOX-1.1) (M6-12) (M6); Homeobox protein M6-12; homeobox protein R5; HOX1; HOX1.1; hox-1.1; HOX1A; Hox1r5; Hoxa7; Hoxa-7; Hoxa9; M6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3204 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P31268 | |
HOXA7 | |
Recombinant Human HOXA7 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only