missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXC4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | HOXC4 |
---|---|
Concentration | 0.32 mg/mL |
Applications | Western Blot, Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16832904
|
Proteintech
14321-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16822904
|
Proteintech
14321-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.Specifications
HOXC4 | |
Western Blot, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
P09017, P18865, Q08624 | |
15423, 24459, 3221 | |
HOXC4 Fusion Protein Ag5563 | |
Primary | |
-20°C | |
HOXC4 |
0.32 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
cp19, homeobox C4, Homeobox protein CP19, Homeobox protein Hox 3E, Homeobox protein Hox C4, HOX3, HOX3E, HOXC4 | |
HOXC4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |