missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXD8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18588-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXD genes located in a cluster on chromosome 2. Deletions that remove the entire HOXD gene cluster or the 5′ end of this cluster have been associated with severe limb and genital abnormalities. In addition to effects during embryogenesis, this particular gene may also play a role in adult urogenital tract function.Specifications
HOXD8 | |
Polyclonal | |
Unconjugated | |
HOXD8 | |
homeobox D8, Homeobox protein Hox 4E, Homeobox protein Hox 5.4, Homeobox protein Hox D8, HOX4, HOX4E, HOX5.4, HOXD8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100912936, 15437, 3234 | |
-20°C | |
Liquid |
Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P13378, P23463 | |
HOXD8 | |
HOXD8 Fusion Protein Ag13245 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |