missing translation for 'onlineSavingsMsg'
Learn More

HPCAL4 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12470-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16880194

  • 3313.85 SEK / 150µL
Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

May be involved in the calcium-dependent regulation of rhodopsin phosphorylation.
Specifications

Specifications

HPCAL4
0.26 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9UM19
hippocalcin like 4, Hippocalcin like protein 4, HLP4, HPCAL4
Rabbit
IgG
150 μL
-20°C
Primary
51440
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
HPCAL4
Liquid
HPCAL4
HPCAL4 Fusion Protein Ag3153
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers