missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPx2 Monoclonal Antibody (HIC1-1C10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516133
Additional Details : Weight : 0.10000kg
Description
Stem cells offer a therapeutic promise for many diseases. In the case of diabetes, stem cells offer the promise that beta cells may be generated ex vivo for the possible transplantation and cure of Type I diabetes (Couri CE, et al.). For this to happen, stem cells will need to be isolated and expanded, they will need to be differentiated toward the beta cell lineage, and the differentiated cell progeny used for transplantation may need to be isolated from complex cell mixtures following differentiation. Appropriate markers targeting cell surface molecules on developing and mature pancreatic cells will be required for such research.Specifications
HPx2 | |
Monoclonal | |
1 mg/mL | |
tissue culture supernatant with 0.05% sodium azide | |
Human pancreatic enriched exocrine cells | |
RUO | |
Human | |
Antibody | |
IgM |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen) | |
HIC1-1C10 | |
Unconjugated | |
Mouse | |
100 μL | |
Primary | |
4° C, do not freeze | |
Liquid |