missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
HRAS Antibody (Y13-238) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP3-20119-0.025mg
This item is not returnable.
View return policy
Description
HRAS Monoclonal antibody specifically detects HRAS in Human, Mouse samples. It is validated for Immunoprecipitation
Specifications
| HRAS | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Rat | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation | |
| Y13-238 | |
| Unconjugated | |
| C-BAS/HAS;C-HA-RAS1;C-H-RAS;CTLO;GTPase HRas;HAMSV;HRAS1;H-RASIDX;p21ras;RASH1 | |
| Injection of rats subcutaneously with Ha-MuSV transformed NRK cells and screening of sera of individual animals to identify those with the strongest immune response against p21. Spleens were removed from the immunised animals and cell fusion of immune lymphocytes with rat myeloma cell line 210RCY3 Ag 1.2.3 (Y3) was carried out. Supernatants were harvested and assayed for antibodies to p21 by GDP binding assay or immunofluorescence on Ha-SV-transformed MDCK cells, followed by testing for the ability of the antibodies to immunoprecipitate p21. | |
| 0.025 mg | |
| Cell Cycle and Replication, Metastasis, mTOR Pathway | |
| 3265 | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
| IgG2a λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction