Learn More
HRD1/SYVN1 Mouse anti-Human, Unconjugated, Clone: 2C8A7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67488-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The HRD1/SYVN1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HRD1/SYVN1. This antibody recognizes human, mouse, rat antigen. The HRD1/SYVN1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
HRD1/SYVN1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
2C8A7 | |
Unconjugated | |
150 μL | |
84447 | |
IgG2b |