missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRG Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | HRG |
---|---|
Concentration | 0.36 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16884765
|
Proteintech
26252-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16874765
|
Proteintech
26252-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
HRG contains two cystatin-like domains and is located in plasma and platelets. The physiological function has not been determined but it is known that the protein binds heme, dyes and divalent metal ions. It can inhibit rosette formation and interacts with heparin, thrombospondin and plasminogen. Two of the protein′s effects, the inhibition of fibrinolysis and the reduction of inhibition of coagulation, indicate a potential prothrombotic effect. Mutations in this gene lead to thrombophilia due to abnormal histidine-rich glycoprotein levels.Specifications
HRG | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P04196 | |
3273 | |
HRG Fusion Protein Ag16278 | |
Primary | |
-20°C | |
HRG |
0.36 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
histidine rich glycoprotein, HPRG, HRG, HRGP | |
HRG | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |