Läs mer
Invitrogen™ HRI Recombinant Superclonal™ Antibody (7HCLC)
Description
This antibody is predicted to react with Monkey, Horse, Mouse Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called Recombinant polyclonal antibody, this product is now rebranded as Recombinant Superclonal™ antibody. The physical product and the performance remain unchanged.
Spécification
Spécification
| Antigen | HRI |
| Användningsområden | Immunoprecipitation, Western Blot, RNA Immunoprecipitation |
| Klassificering | Recombinant Superclonal |
| Klona | 7HCLC |
| Koncentration | 0.5 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 0.09% sodium azide; pH 7.4 |
| Gen | EIF2AK1 |
| Genaccessionsnr. | Q9BQI3 |
| Gene Alias | EIF2AK1; eukaryotic translation initiation factor 2 alpha kinase 1; eukaryotic translation initiation factor 2-alpha kinase 1; HCR; he; heme regulated initiation factor 2 alpha kinase; heme sensitive initiation factor 2a kinase; heme-controlled repressor; Heme-regulated eukaryotic initiation factor eIF-2-alpha kinase; heme-regulated inhibitor; heme-regulated initiation factor 2-alpha kinase; heme-regulated repressor; hemin-sensitive initiation factor 2a kinase; hemin-sensitive initiation factor 2-alpha kinase; Hri; KIAA1369; PRO1362 |
| Afficher plus |
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.