missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ HRSP12 Recombinant Protein

Human full-length ORF recombinant protein with GST-tag at N-terminal

Brand:  Abnova™ H00010247-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16144722

  • 6845.00 SEK / 25µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Encoded by heat-responsive protein 12 gene
  • Molecular weight: 40.81kDa
  • Preparation method: in vitro wheat germ expression system
  • Purification: glutathione sepharose 4 fast flow
  • Storage buffer: 50 mM Tris-HCI, 10 mM reduced glutathione
  • pH=8.0 in elution buffer

Sequence: MSSLIRRVISTAKAPGAIGPYSQAVLVDRTIYISGQIGMDPSSGQLVSGGVAEEAKQALKNMGEILKAAGCDFTNVVKTTVLLADINDFNTVNEIYKQYFKSNFPARAAYQVAALPKGSRIEIEAVAIQGPLTTASL

Best when used within three months from the date of receipt of this protein.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH10280.1
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
40.8
8
Glutathione Sepharose 4 Fast Flow
25 ug
MSSLIRRVISTAKAPGAIGPYSQAVLVDRTIYISGQIGMDPSSGQLVSGGVAEEAKQALKNMGEILKAAGCDFTNVVKTTVLLADINDFNTVNEIYKQYFKSNFPARAAYQVAALPKGSRIEIEAVAIQGPLTTASL
P14.5/PSP/UK114
HRSP12
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
10247
HRSP12 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HRSP12
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers