Läs mer
HSF1 Antibody (4B4), Janelia Fluor™ 646, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-42206JF646
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
HSF1 Monoclonal antibody specifically detects HSF1 in Human, Mouse, Rat, Bovine, Guinea Pig, Hamster, Monkey, Rabbit samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Gel Super Shift Assays
Specifikationer
| HSF1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine, Guinea Pig | |
| Purified |
| ELISA, Immunohistochemistry, Immunoprecipitation, Gel Supershift | |
| 4B4 | |
| ELISA, Immunohistochemistry, Immunoprecipitation, Gel Super Shift Assays | |
| heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1 | |
| Purified recombinant mouse HSF1 protein, epitope mapping to amino acids 425-439 | |
| 0.1 mL | |
| Cancer, DNA Repair, Transcription Factors and Regulators | |
| 3297 | |
| Store at 4°C in the dark. | |
| IgG1 |