missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ HSF4 Monoclonal Antibody (504524)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524217
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
This antibody detects endogenous human and mouse HSF4 in Western blots. Reconstitute at 0.5 mg/mL in sterile PBS.
Heat-shock transcription factors (HSFs) activate heat-shock response genes under conditions of heat or other stresses. HSF4 lacks the carboxyl-terminal hydrophobic repeat which is shared among all vertebrate HSFs and has been suggested to be involved in the negative regulation of DNA binding activity. Two alternatively spliced transcripts encoding distinct isoforms and possessing different transcriptional activity have been described.
Specifikationer
| HSF4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9R0L1, Q9ULV5 | |
| HSF4 | |
| E. coli-derived recombinant human HSF4 Ser309-Pro463. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 504524 | |
| Unconjugated | |
| HSF4 | |
| ATHSF4; AT-HSFB1; binds heat shock DNA elements; cataract; CTM; CTRCT5; DNA binding protein; heat shock factor 4; heat shock factor protein 4; Heat shock transcription factor 4; hHSF4; HSF 4; Hsf4; HSFB1; HSTF 4; HSTF4; ldis1; Marner; mHSF4 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 26386, 3299 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering