missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSFY1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11970-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the heat shock factor (HSF) family of transcriptional activators for heat shock proteins. This gene is a candidate gene for azoospermia, since it localizes to a region of chromosome Y that is sometimes deleted in infertile males. The genome has two identical copies of this gene within a palindromic region; this record represents the more centromeric copy. Alternative splicing results in multiple transcript variants encoding distinct isoforms.Specifications
HSFY1 | |
Polyclonal | |
Unconjugated | |
HSFY1 | |
HSF2 like, HSF2L, HSFY, HSFY1, HSFY2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
86614 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96LI6 | |
HSFY1 | |
HSFY1 Fusion Protein Ag2592 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |