missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPA13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12667-2-AP-20UL
Additional Details : Weight : 0.00010kg
Description
STCH encodes a protein that is a member of the heat shock protein 70 family and is found associated with microsomes. Members of this protein family play a role in the processing of cytosolic and secretory proteins, as well as in the removal of denatured or incorrectly-folded proteins. The encoded protein contains an ATPase domain and has been shown to associate with a ubiquitin-like protein.Specifications
HSPA13 | |
Polyclonal | |
Unconjugated | |
HSPA13 | |
Heat shock 70 kDa protein 13, HSPA13, STCH | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
110920, 29734, 6782 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35162, P48723, Q8BM72 | |
Hspa13 | |
HSPA13 Fusion Protein Ag3349 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |