missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPG2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19675-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The antibody is specific to HSPG2.
Perlecan is a major heparan-sulfate proteoglycan (HSPG) found within all basement membranes and cell surfaces. Because of its strategic location and ability to store and protect growth factors, perlecan has been strongly implicated in the control of tumor cell growth and metastatic behavior. Perlecan possesses angiogenic and growth-promoting attributes primarily by acting as a coreceptor for basic fibroblast growth factor (FGF-2). Suppression of perlecan causes substantial inhibition of neoplastic growth and neovascularization. Thus, perlecan is a potent inducer of neoplasm growth and angiogenesis in vivo and therapeutic interventions targeting this key modulator of tumor progression may improve neoplastic treatment.Specifications
HSPG2 | |
Polyclonal | |
Unconjugated | |
HSPG2 | |
heparan sulfate proteoglycan 2, HSPG, HSPG2, Perlecan, PLC, PRCAN, SJA, SJS, SJS1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15530, 313641, 3339 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P98160 | |
Hspg2 | |
Peptide | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |