missing translation for 'onlineSavingsMsg'
Learn More

HTR1A Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 55302-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16826795

  • 3313.85 SEK / 150µL
Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This is one of the several different receptors for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by G proteins that inhibit adenylate cyclase activity.
Specifications

Specifications

HTR1A
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P08908
5 HT 1A, 5 HT1A, 5HT1a, ADRB2RL1, ADRBRL1, G 21, HTR1A, Serotonin receptor 1A
Rabbit
IgG
150 μL
-20°C
Primary
3350
Immunohistochemistry (Paraffin)
Unconjugated
HTR1A
Liquid
HTR1A
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers