missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ HTR1D (Human) Recombinant Protein
Human HTR1D full-length ORF ( AAH07720.1, 1 a.a. - 377 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00003352-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSPLNQSAEGLPQEASNRSLNATETSEAWDPRTLQALKISLAVVLSVITLATVLSNAFVLTTILLTRKLHTPANYLIGSLATTDLLVSILVMPISIAYTITHTWNFGQILCDIWLSSDITCCTASILHLCVIALDRYWAITDALEYSKRRTAGHAATMIAIVWAISICISIPPLFWRQAKAQEEMPDCLVNTSQISYTIYSTCGAFYIPSVLLIILYGRIYRAARNRILNPPSLYGKRFTTAHLITGSAGSSLCSLNSSLHEGHSHSAGSPLFFNHVKIKLADSALERERISAARERKATKILGIILGAFIICWLPFFVVSLVLPICRDSCWIHPALFDFFTWLGYLNSLINPIIYTVFNEEFRQAFQKIVPFRKAS
Specifications
AAH07720.1 | |
5-hydroxytryptamine (serotonin) receptor 1D | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HTR1D | |
GST |
3352 | |
Wheat germ expression system | |
25 μg | |
5-HT1D, HT1DA, HTR1DA, HTRL, RDC4 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |