missing translation for 'onlineSavingsMsg'
Learn More

HTR3A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10443-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16876833

  • 3313.85 SEK / 150µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The product of this gene belongs to the ligand-gated ion channel receptor superfamily. This gene encodes subunit A of the type 3 receptor for 5-hydroxytryptamine, a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor causes fast, depolarizing responses in neurons after activation. It appears that the heteromeric combination of A and B subunits is necessary to provide the full functional features of this receptor, since either subunit alone results in receptors with very low conductance and response amplitude. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications

Specifications

HTR3A
0.15 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P23979, P35563, P46098
5 HT 3, 5 HT3 A, 5 HT3A, 5 HT3R, 5 hydroxytryptamine receptor 3, 5HT3R, HTR3, HTR3A, Serotonin receptor 3A
Rabbit
IgG
150 μL
-20°C
Primary
15561, 3359, 79246
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
HTR3A
Liquid
HTR3A
HTR3A Fusion Protein Ag0289
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers