Learn More
HTR6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23561-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This is one of the several different receptors for 5-hydroxytryptamine, a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by G proteins that stimulate adenylate cyclase. It has a high affinity for tricyclic psychotropic drugs.Specifications
HTR6 | |
Polyclonal | |
Unconjugated | |
HTR6 | |
5 HT 6, 5 HT6, 5 HT6R, 5 hydroxytryptamine receptor 6, HTR6, Serotonin receptor 6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3362 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P50406 | |
HTR6 | |
HTR6 Fusion Protein Ag15396 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |