missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ HTRA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121488
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
HtrA1, a 51KDa protein, is a member of highly conserved mammalian HtrA serine protease family. It is a human homolog of bacterial HtrA and consists of a secretory signal sequence, an IGFBP-binding domain and a Kazal-type serine-protease inhibitor domain at the N-terminus and a highly conserved trypsin-like serine/protease domain and a PDZ domain at the C-terminus. It plays a major role in the regulation of cell growth. HtrA1 inhibits signaling mediated by TGF-beta family members and regulates many physiological processes. HtrA1 is up-regulated in placenta during pregnancy and plays an important role in implantation and placental physiology. It regulates the availability of IGFs by cleaving IGF-binding proteins. It acts as a tumor suppressor and is down-regulated in ovarian cancers and melanomas. Its role has been implicated in osteoarthritis suggesting a therapeutic intervention. Northern blot analysis detected high expression in placenta.
Specifikationer
| HTRA1 | |
| Polyclonal | |
| Unconjugated | |
| AI429470; ARMD7; CADASIL2; CARASIL; High-temperature requirement A serine peptidase 1; HTRA; HtrA serine peptidase 1; Htra1; IGF binding; IGFBP5-protease; insulin-like growth factor binding protein 5 protease; L56; ORF480; protease serine 11; protease, serine, 11; protease, serine, 11 (IGF binding); PRSS11; RSPP11; serine protease; serine protease 11; Serine protease HTRA1; similar to bacterial htrA and degP | |
| Lys304-Lys442 (Accession P0C0V0), with N-terminal His Tag. | |
| 20 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering