missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HTRA4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | HTRA4 |
---|---|
Concentration | 0.22 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16832584
|
Proteintech
14095-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16822584
|
Proteintech
14095-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Serine protease.Specifications
HTRA4 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
A2RT60, D3ZKF5, P83105 | |
203100, 306564, 330723 | |
HTRA4 Fusion Protein Ag5227 | |
Primary | |
-20°C | |
HTRA4 |
0.22 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
HtrA serine peptidase 4, HTRA4, Probable serine protease HTRA4 | |
Htra4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |