missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HTRA4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14095-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Serine protease.Specifications
HTRA4 | |
Polyclonal | |
Unconjugated | |
HTRA4 | |
HtrA serine peptidase 4, HTRA4, Probable serine protease HTRA4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
203100, 306564, 330723 | |
-20°C | |
Liquid |
Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A2RT60, D3ZKF5, P83105 | |
Htra4 | |
HTRA4 Fusion Protein Ag5227 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |