missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human 40787 Full-length ORF (AAH12161, 1 a.a. - 367 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001731-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16194041

  • 6845.00 SEK / 25µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is a member of the septin family of GTPases. Members of this family are required for cytokinesis. This gene encodes a protein associated with the tau-based paired helical filament core, and may contribute to the formation of neurofibrillary tangles in Alzheimer's disease. [provided by RefSeq]

Sequence: MDKEYVGFAALPNQLHRKSVKKGFDFTLMVAGESGLGKSTLINSLFLTNLYEDRQVPEASARLTQTLAIERRGVEIEEGGVKVKLTLVDTPGFGDSVDCSDCWLPVVKFIEEQFEQYLRDESGLNRKNIQDSRVHCCLYFISPFGRGLRPLDVAFLRAVHEKVNIIPVIGKADALMPQETQALKQKIRDQLKEEEIHIYQFPECDSDEDEDFKRQDAEMKESIPFAVVGSCEVVRDGGNRPVRGRRYSWGTVEVENPHHCDFLNLRRMLVQTHLQDLKEVTHDLLYEGYRARRLQSLARPGARDRASRSKLSRQSATEIPLPMLPLADTEKLIREKDEELRRMQEMLEKMQAQMQRSQAQGEQSDAL
Specifications

Specifications

AAH12161
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
66.11kDa
Glutathione Sepharose 4 Fast Flow
25μg
RUO
SEPT1
Wheat Germ (in vitro)
GST Tag
Liquid
Protein Array, ELISA, Western Blot
1731
Human 40787 Full-length ORF (AAH12161, 1 a.a. - 367 a.a.) Recombinant Protein with GST-tag at N-terminal
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at −80°C. Aliquot to avoid repeated freezing and thawing.
DIFF6, LARP, MGC20394, PNUTL3, SEP1
SEPT1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers