missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AASDH Full-length ORF (AAH15096.1, 1 a.a. - 134 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00132949-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16177983

  • 3343.96 SEK / 10µg

Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MTLQELVHKAASCYMDRVAVCFDECNNQLPVYYTYKTVVNAASELSNFLLLHCDFQGIREIGLYCQPGIDLPSWILGILQVPAAYVPIEPDSPPSLSTHFMKKCNLKYILVEKKQINVSLDVSIVFCLYYIYFN
Specifications

Specifications

AAH15096.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
41.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
AASDH
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
132949
AASDH (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ACSF4/LYS2/NRPS1098/NRPS998
AASDH
Yes
Videos
SDS
Documents
Certificates