Learn More
Abnova™ Human AASDHPPT Full-length ORF (AAH15470, 1 a.a. - 309 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00060496-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is similar to Saccharomyces cerevisiae LYS5, which is required for the activation of the alpha-aminoadipate dehydrogenase in the biosynthetic pathway of lysine. Yeast alpha-aminoadipate dehydrogenase converts alpha-biosynthetic-aminoadipate semialdehyde to alpha-aminoadipate. It has been suggested that defects in the human gene result in pipecolic acidemia. [provided by RefSeq]
Sequence: MVFPAKRFCLVPSMEGVRWAFSCGTWLPSRAEWLLAVRSIQPEEKERIGQFVFARDAKAAMAGRLMIRKLVAEKLNIPWNHIRLQRTARGKPVLAKDSSNPYPNFNFNISHQGDYAVLAAEPELQVGIDIMKTSFPGRGSIPEFFHIMKRKFTNKEWETIRSFKDEWTQLDMFYRNWALKESFIKAIGAGLGFELQRLEFDLSPLNLDIGQVYKETRLFLDGEEEKEWAFEESKIDEHHFVAVALRKPDGSRHQDVPSQDDSKPTQRQFTILNFNDLMSSAVPMTPEDPSFWDCFCFTEEIPIRNGTKSSpecifications
AAH15470 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
59.73kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
AASDHPPT | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
60496 | |
AASDHPPT (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AASD-PPT/CGI-80/DKFZp566E2346/LYS2/LYS5 | |
AASDHPPT | |
Recombinant | |
wheat germ expression system |