Learn More
Abnova™ Human ABCC13 Full-length ORF (ENSP00000345983, 1 a.a. - 169 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00150000-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the superfamily of genes encoding ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, and White). This family member is part of the MRP subfamily, which is involved in multi-drug resistance, but the human locus is now thought to be a pseudogene incapable of encoding a functional ABC protein. Alternative splicing results in multiple transcript variants; however, not all variants have been fully described. [provided by RefSeq]
Sequence: MLSSTQNAGGSYQRVRGALDTQKCSPEKSASFFSKVTYSWFSRVITLGYKRPLEREDLFELKESDSFCTACPIFEKQWRKEVLRNQERQKVKVSCYKEAHIKKPSLLYALWNTFKSILIQVALFKVFADILSFTSPLIMNYTRKVNYLMGLPCENQKITSYSQASGRDSSpecifications
ENSP00000345983 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
45.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
ABCC13 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
150000 | |
ABCC13 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C21orf73/PRED6 | |
ABCC13 | |
Recombinant | |
wheat germ expression system |