Learn More
Abnova™ Human ABCD1 Full-length ORF (NP_000024.2, 1 a.a. - 745 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_000024.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 215 |
Molecular Weight (g/mol) | 109.3kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16181441
|
Abnova™
H00000215-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16171441
|
Abnova™
H00000215-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the ALD subfamily, which is involved in peroxisomal import of fatty acids and/or fatty acyl-CoAs in the organelle. All known peroxisomal ABC transporters are half transporters which require a partner half transporter molecule to form a functional homodimeric or heterodimeric transporter. This peroxisomal membrane protein is likely involved in the peroxisomal transport or catabolism of very long chain fatty acids. Defects in this gene have been identified as the underlying cause of adrenoleukodystrophy, an X-chromosome recessively inherited demyelinating disorder of the nervous system. [provided by RefSeq]
Sequence: MPVLSRPRPWRGNTLKRTAVLLALAAYGAHKVYPLVRQCLAPARGLQAPAGEPTQEASGVAAAKAGMNRVFLQRLLWLLRLLFPRVLCRETGLLALHSAALVSRTFLSVYVARLDGRLARCIVRKDPRAFGWQLLQWLLIALPATFVNSAIRYLEGQLALSFRSRLVAHAYRLYFSQQTYYRVSNMDGRLRNPDQSLTEDVVAFAASVAHLYSNLTKPLLDVAVTSYTLLRAARSRGAGTAWPSAIAGLVVFLTANVLRAFSPKFGELVAEEARRKGELRYMHSRVVANSEEIAFYGGHEVELALLQRSYQDLASQINLILLERLWYVMLEQFLMKYVWSASGLLMVAVPIITATGYSESDAEAVKKAALEKKEEELVSERTEAFTIARNLLTAAADAIERIMSSYKEVTELAGYTARVHEMFQVFEDVQRCHFKRPRELEDAQAGSGTIGRSGVRVEGPLKIRGQVVDVEQGIICENIPIVTPSGEVVVASLNIRVEEGMHLLITGPNGCGKSSLFRILGGLWPTYGGVLYKPPPQRMFYIPQRPYMSVGSLRDQVIYPDSVEDMQRKGYSEQDLEAILDVVHLHHILQREGGWEAMCDWKDVLSGGEKQRIGMARMFYHRPKYALLDECTSAVSIDVEGKIFQAAKDAGIALLSITHRPSLWKYHTHLLQFDGEGGWKFEKLDSAARLSLTEEKQRLEQQLAGIPKMQRRLQELCQILGEAVAPAHVPAPSPQGPGGLQGASTSpecifications
NP_000024.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
109.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ABC42/ALD/ALDP/AMN | |
ABCD1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
215 | |
ABCD1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
ABCD1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |