Learn More
Abnova™ Human ABI1 Partial ORF (NP_001012770, 185 a.a. - 238 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010006-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene has been found to form a complex with EPS8 and SOS1, and is thought to be involved in the transduction of signals from Ras to Rac. In addition, the encoded protein may play a role in the regulation of EGF-induced Erk pathway activation as well as cytoskeletal reorganization and EGFR signaling. Several transcript variants encoding multiple isoforms have been found for this gene. [provided by RefSeq]
Sequence: GTLGRNTPYKTLEPVKPPTVPNDYMTSPARLGSQHSPGRTASLNQRPRTHSGSSSpecifications
NP_001012770 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
31.68kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GTLGRNTPYKTLEPVKPPTVPNDYMTSPARLGSQHSPGRTASLNQRPRTHSGSS | |
RUO | |
ABI1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10006 | |
ABI1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ABI-1/E3B1/NAP1BP/SSH3BP/SSH3BP1 | |
ABI1 | |
Recombinant | |
wheat germ expression system |