Learn More
Abnova™ Human ACAA2 Full-length ORF (AAH01918.1, 1 a.a. - 397 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010449-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The encoded protein catalyzes the last step of the mitochondrial fatty acid beta-oxidation spiral. Unlike most mitochondrial matrix proteins, it contains a non-cleavable amino-terminal targeting signal. [provided by RefSeq]
Sequence: MALLRGVFVVAAKRTPFGAYGGLLKDFTATDLSEFAAKAALSAGKVSPETVDSVIMGNVLQSSSDAIYLARHVGLRVGIPKETPALTINRLCGSGFQSIVNGCQEICVKEAEVVLCGGTESMSQAPYCVRNVRFGTKLGSDIKLEDSLWVSLTDQHVQLPMAMTAENLAVKHKISREECDKYALQSQQRWKAANDAGYFNDEMAPIEVKTKKGKQTMQVDEHARPQTTLEQLQKLPPVFKKDGTVTAGNASGVADGAGAVIIASEDAVKKHNFTPLARIVGYFVSGCDPSIMGIGPVPAISGALKKAGLSLKDMDLVEVNEAFAPQYLAVERSLDLDISKTNVNGGAIALGHPLGGSGSRITAHLVHELRRRGGKYAVGSACIGGGQGIAVIIQSTASpecifications
AAH01918.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
68.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
ACAA2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10449 | |
ACAA2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DSAEC/FLJ35992/FLJ95265 | |
ACAA2 | |
Recombinant | |
wheat germ expression system |