Learn More
Abnova™ Human ACAD8 Partial ORF (NP_055199, 306 a.a. - 415 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00027034-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Isobutyryl-CoA dehydrogenase (EC 1.3.99), encoded by the ACAD8 gene, catalyzes the third step of the degradation of the branched chain amino acid valine (Nguyen et al., 2002 [PubMed 12359132]).[supplied by OMIM]
Sequence: GEPLASNQYLQFTLADMATRLVAARLMVRNAAVALQEERKDAVALCSMAKLFATDECFAICNQALQMHGGYGYLKDYAVQQYVRDSRVHQILEGSNEVMRILISRSLLQESpecifications
NP_055199 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GEPLASNQYLQFTLADMATRLVAARLMVRNAAVALQEERKDAVALCSMAKLFATDECFAICNQALQMHGGYGYLKDYAVQQYVRDSRVHQILEGSNEVMRILISRSLLQE | |
RUO | |
ACAD8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
27034 | |
ACAD8 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACAD-8/FLJ22590 | |
ACAD8 | |
Recombinant | |
wheat germ expression system |