missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ACAD8 Partial ORF (NP_055199, 306 a.a. - 415 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00027034-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16185636

  • 4515.00 SEK / 10µg
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Isobutyryl-CoA dehydrogenase (EC 1.3.99), encoded by the ACAD8 gene, catalyzes the third step of the degradation of the branched chain amino acid valine (Nguyen et al., 2002 [PubMed 12359132]).[supplied by OMIM]

Sequence: GEPLASNQYLQFTLADMATRLVAARLMVRNAAVALQEERKDAVALCSMAKLFATDECFAICNQALQMHGGYGYLKDYAVQQYVRDSRVHQILEGSNEVMRILISRSLLQE
Specifications

Specifications

NP_055199
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GEPLASNQYLQFTLADMATRLVAARLMVRNAAVALQEERKDAVALCSMAKLFATDECFAICNQALQMHGGYGYLKDYAVQQYVRDSRVHQILEGSNEVMRILISRSLLQE
RUO
ACAD8
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
27034
ACAD8 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ACAD-8/FLJ22590
ACAD8
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers