missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ACAD9 Full-length ORF (NP_054768.2, 1 a.a. - 621 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_054768.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 28976 |
Molecular Weight (g/mol) | 95.2kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16178902
|
Abnova™
H00028976-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16168902
|
Abnova™
H00028976-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Mitochondrial fatty acid beta-oxidation is one of the main energy-producing metabolic pathways in eukaryotes. Acyl-CoA dehydrogenases (ACADs; EC 1.3.99.13) are mitochondrial enzymes that catalyze the initial rate-limiting step in the beta-oxidation of fatty acyl-CoA. ACAD9 belongs to a group of ACADs that act on fatty acids containing 14 to 20 carbons (Zhang et al., 2002 [PubMed 12359260]).[supplied by OMIM]
Sequence: MSGCGLFLRTTAAARACRGLVVSTANRRLLRTSPPVRAFAKELFLGKIKKKEVFPFPEVSQDELNEINQFLGPVEKFFTEEVDSRKIDQEGKIPDETLEKLKSLGLFGLQVPEEYGGLGFSNTMYSRLGEIISMDGSITVTLAAHQAIGLKGIILAGTEEQKAKYLPKLASGEHIAAFCLTEPASGSDAASIRSRATLSEDKKHYILNGSKVWITNGGLANIFTVFAKTEVVDSDGSVKDKITAFIVERDFGGVTNGKPEDKLGIRGSNTCEVHFENTKIPVENILGEVGDGFKVAMNILNSGRFSMGSVVAGLLKRLIEMTAEYACTRKQFNKRLSEFGLIQEKFALMAQKAYVMESMTYLTAGMLDQPGFPDCSIEAAMVKVFSSEAAWQCVSEALQILGGLGYTRDYPYERILRDTRILLIFEGTNEILRMYIALTGLQHAGRILTTRIHELKQAKVSTVMDTVGRRLRDSLGRTVDLGLTGNHGVVHPSLADSANKFEENTYCFGRTVETLLLRFGKTIMEEQLVLKRVANILINLYGMTAVLSRASRSIRIGLRNHDHEVLLANTFCVEAYLQNLFSLSQLDKYAPENLDEQIKKVSQQILEKRAYICAHPLDRTCSpecifications
NP_054768.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
95.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACAD-9/FLJ23533/MGC14452/NPD002 | |
ACAD9 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
28976 | |
ACAD9 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
ACAD9 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |