Learn More
Abnova™ Human ACCN2 Partial ORF (NP_064423.2, 96 a.a. - 185 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_064423.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 41 |
Molecular Weight (g/mol) | 35.64kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16194264
|
Abnova™
H00000041-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16184264
|
Abnova™
H00000041-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the degenerin/epithelial sodium channel (DEG/ENaC) superfamily. The members of this family are amiloride-sensitive sodium channels that contain intracellular N and C termini, 2 hydrophobic transmembrane regions, and a large extracellular loop, which has many cysteine residues with conserved spacing. The member encoded by this gene is expressed in most if not all brain neurons, and it may be an ion channel subunit; however, its function as an ion channel remains unknown. Alternative splicing of this gene generates 2 transcript products. [provided by RefSeq]
Sequence: NEFRFSQVSKNDLYHAGELLALLNNRYEIPDTQMADEKQLEILQDKANFRSFKPKPFNMREFYDRAGHDIRDMLLSCHFRGEVCSAEDFKSpecifications
NP_064423.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ASIC/ASIC1/ASIC1A/BNaC2/hBNaC2 | |
ACCN2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
41 | |
ACCN2 (Human) Recombinant Protein (Q01) | |
NEFRFSQVSKNDLYHAGELLALLNNRYEIPDTQMADEKQLEILQDKANFRSFKPKPFNMREFYDRAGHDIRDMLLSCHFRGEVCSAEDFK | |
RUO | |
ACCN2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |