missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ACOX2 Full-length ORF (NP_003491.1, 1 a.a. - 681 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_003491.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8309 |
Molecular Weight (g/mol) | 103.2kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16144738
|
Abnova™
H00008309-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16134738
|
Abnova™
H00008309-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The product of this gene belongs to the acyl-CoA oxidase family. It encodes the branched-chain acyl-CoA oxidase which is involved in the degradation of long branched fatty acids and bile acid intermediates in peroxisomes. Deficiency of this enzyme results in the accumulation of branched fatty acids and bile acid intermediates, and may lead to Zellweger syndrome, severe mental retardation, and death in children. [provided by RefSeq]
Sequence: MGSPVHRVSLGDTWSRQMHPDIESERYMQSFDVERLTNILDGGAQNTALRRKVESIIHSYPEFSCKDNYFMTQNERYKAAMRRAFHIRLIARRLGWLEDGRELGYAYRALSGDVALNIHRVFVRALRSLGSEEQIAKWDPLCKNIQIIATYAQTELGHGTYLQGLETEATYDAATQEFVIHSPTLTATKWWPGDLGRSATHALVQAQLICSGARRGMHAFIVPIRSLQDHTPLPGIIIGDIGPKMDFDQTDNGFLQLNHVRVPRENMLSRFAQVLPDGTYVKLGTAQSNYLPMVVVRVELLSGEILPILQKACVIAMRYSVIRRQSRLRPSDPEAKVLDYQTQQQKLFPQLAISYAFHFLAVSLLEFFQHSYTAILNQDFSFLPELHALSTGMKAMMSEFCTQGAEMCRRACGGHGYSKLSGLPSLVTKLSASCTYEGENTVLYLQVARFLVKSYLQTQMSPGSTPQRSLSPSVAYLTAPDLARCPAQRAADFLCPELYTTAWAHVAVRLIKDSVQHLQTLTQSGADQHEAWNQTTVIHLQAAKVHCYYVTVKGFTEALEKLENEPAIQQVLKRLCDLHAIHGILTNSGDFLHDAFLSGAQVDMARTAYLDLLRLIRKDAILLTDAFDFTDQCLNSALGCYDGNVYERLFQWAQKSPTNTQENPAYEEYIRPLLQSWRSKLSpecifications
NP_003491.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
103.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BCOX/BRCACOX/BRCOX/THCCox | |
ACOX2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8309 | |
ACOX2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
ACOX2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |