missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ACSBG1 Full-length ORF (NP_055977.3, 1 a.a. - 724 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00023205-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16137052

  • 6025.00 SEK / 25µg
Estimated Shipment: 03-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene possesses long-chain acyl-CoA synthetase activity. It is thought to play a central role in brain very long-chain fatty acids metabolism and myelinogenesis. [provided by RefSeq]

Sequence: MPRNSGAGYGCPHGDPSMLDSRETPQESRQDMIVRTTQEKLKTSSLTDRQPLSKESLNHALELSVPEKVNNAQWDAPEEALWTTRADGRVRLRIDPSCPQLPYTVHRMFYEALDKYGDLIALGFKRQDKWEHISYSQYYLLARRAAKGFLKLGLKQAHSVAILGFNSPEWFFSAVGTVFAGGIVTGIYTTSSPEACQYIAYDCCANVIMVDTQKQLEKILKIWKQLPHLKAVVIYKEPPPNKMANVYTMEEFMELGNEVPEEALDAIIDTQQPNQCCVLVYTSGTTGNPKGVMLSQDNITWTARYGSQAGDIRPAEVQQEVVVSYLPLSHIAAQIYDLWTGIQWGAQVCFAEPDALKGSLVNTLREVEPTSHMGVPRVWEKIMERIQEVAAQSGFIRRKMLLWAMSVTLEQNLTCPGSDLKPFTTRLADYLVLAKVRQALGFAKCQKNFYGAAPMMAETQHFFLGLNIRLYAGYGLSETSGPHFMSSPYNYRLYSSGKLVPGCRVKLVNQDAEGIGEICLWGRTIFMGYLNMEDKTCEAIDEEGWLHTGDAGRLDADGFLYITGRLKELIITAGGENVPPVPIEEAVKMELPIISNAMLIGDQRKFLSMLLTLKCTLDPDTSDQTDNLTEQAMEFCQRVGSRATTVSEIIEKKDEAVYQAIEEGIRRVNMNAAARPYHIQKWAILERDFSISGGELGPTMKLKRLTVLEKYKGIIDSFYQEQKM
Specifications

Specifications

NP_055977.3
Liquid
23205
ACSBG1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BG1/BGM/FLJ30320/GR-LACS/KIAA0631/MGC14352/hBG1/hsBG
ACSBG1
Yes
wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
107.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
ACSBG1
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers