Learn More
Abnova™ Human ACTR1B Full-length ORF (NP_005726.1, 1 a.a. - 376 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010120-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a 42.3 kD subunit of dynactin, a macromolecular complex consisting of 10 subunits ranging in size from 22 to 150 kD. Dynactin binds to both microtubules and cytoplasmic dynein and is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit, like ACTR1A, is an actin-related protein. These two proteins, which are of equal length and share 90% amino acid identity, are present in a constant ratio of approximately 1:15 in the dynactin complex. [provided by RefSeq]
Sequence: MESYDIIANQPVVIDNGSGVIKAGFAGDQIPKYCFPNYVGRPKHMRVMAGALEGDLFIGPKAEEHRGLLTIRYPMEHGVVRDWNDMERIWQYVYSKDQLQTFSEEHPVLLTEAPLNPSKNREKAAEVFFETFNVPALFISMQAVLSLYATGRTTGVVLDSGDGVTHAVPIYEGFAMPHSIMRVDIAGRDVSRYLRLLLRKEGVDFHTSAEFEVVRTIKERACYLSINPQKDEALETEKVQYTLPDGSTLDVGPARFRAPELLFQPDLVGDESEGLHEVVAFAIHKSDMDLRRTLFANIVLSGGSTLFKGFGDRLLSEVKKLAPKDIKIKISAPQERLYSTWIGGSILASLDTFKKMWVSKKEYEEDGSRAIHRKTFSpecifications
NP_005726.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
68.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
ACTR1B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10120 | |
ACTR1B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARP1B/CTRN2/PC3 | |
ACTR1B | |
Recombinant | |
wheat germ expression system |