Learn More
Abnova™ Human ADAM22 Full-length ORF (AAH36029.1, 1 a.a. - 342 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH36029.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 53616 |
Molecular Weight (g/mol) | 65.9kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16130413
|
Abnova™
H00053616-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16120413
|
Abnova™
H00053616-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This gene is highly expressed in the brain and may function as an integrin ligand in the brain. Alternative splicing results in several transcript variants. [provided by RefSeq]
Sequence: MQAAVAVSVPFLLLCVLGTCPPARCGQAGDASLMELEKRKENRFVERQSIVPLRLIYRSGGEDESRHDALDTRVRGDVGGPQLTHVDQASFQVDAFGTSFILDVVLNHDLLSSEYIERHIEHGGKTVEVKGGEHCYYQGHIRGNPDSFVALSTCHGLHGMFYDGNHTYLIEPEENDTTQEDFHFHSVYKSRLFEFSLDDLPSEFQQVNITPSKFILKPRPKRSKRQLRRYPRNVEEETKYIELMIVNDHLMFKKHRLSVVHTNTYAKSVVNMADLIYKDQLKTRIVLVAMETWATDNKFAISENPLITLREFMKYRRDFIKEKSDAVHLFSYVTSVMMYYFFSpecifications
AAH36029.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
65.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MDC2/MGC149832 | |
ADAM22 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
53616 | |
ADAM22 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
ADAM22 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |