Learn More
Abnova™ Human ADAM33 Partial ORF (NP_079496, 551 a.a. - 650 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_079496 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 80332 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16120277
|
Abnova™
H00080332-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16130277
|
Abnova™
H00080332-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This protein is a type I transmembrane protein implicated in asthma and bronchial hyperresponsiveness. Alternative splicing of this gene results in two transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: AHGNCGQDSEGHFLPCAGRDALCGKLQCQGGKPSLLAPHMVPVDSTVHLDGQEVTCRGALALPSAQLDLLGLGLVEPGTQCGPRMVCQSRRCRKNAFQELSpecifications
NP_079496 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C20orf153/DJ964F7.1/DKFZp434K0521/FLJ35308/FLJ36751/MGC149823/MGC71889 | |
ADAM33 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
80332 | |
ADAM33 (Human) Recombinant Protein (Q01) | |
AHGNCGQDSEGHFLPCAGRDALCGKLQCQGGKPSLLAPHMVPVDSTVHLDGQEVTCRGALALPSAQLDLLGLGLVEPGTQCGPRMVCQSRRCRKNAFQEL | |
RUO | |
ADAM33 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |