missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ADAM7 Full-length ORF (AAI48736.1, 1 a.a. - 754 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008756-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16182852

  • 4515.00 SEK / 10µg
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The ADAM family is composed of zinc-binding proteins that can function as adhesion proteins and/or endopeptidases. They are involved in a number of biologic processes, including fertilization, neurogenesis, muscle development, and immune response.[supplied by OMIM]

Sequence: MLPGCIFLMILLIPQVKEKFILGVEGQQLVRPKKLPLIQKRDTGHTHDDDILKTYEEELLYEIKLNRKTLVLHLLRSREFLGSNYSETFYSMKGEAFTRHPQIMDHCFYQGSIVHEYDSAASISTCNGLRGFFRINDQRYLIEPVKYSDEGEHLVFKYNLRVPYGANYSCTELNFTRKTVPGDNESEEDSKIKGIHDEKYVELFIVADDTVYRRNGHPHNKLRNRIWGMVNFVNMIYKTLNIHVTLVGIEIWTHEDKIELYSNIETTLLRFSFWQEKILKTRKDFDHVVLLSGKWLYSHVQGISYPGGMCLPYYSTSIIKDLLPDTNIIANRMAHQLGHNLGMQHDEFPCTCPSGKCVMDSDGSIPALKFSKCSQNQYHQYLKDYKPTCMLNIPFPYNFHDFQFCGNKKLDEGEECDCGPAQECTNPCCDAHTCVLKPGFTCAEGECCESCQIKKAGSICRPAKDECDFPEMCTGHSPACPKDQFRVNGFPCKNSEGYCFMGKCPTREDQCSELFDDEAIESHDICYKMNTKGNKFGYCKNKENRFLPCEEKDVRCGKIYCTGGELSSLLGEDKTYHLKDPQKNATVKCKTIFLYHDSTDIGLVASGTKCGEGMVCNNGECLNMEKVYISTNCPSQCNENPVDGHGLQCHCEEGQAPVACEETLHVTNITILVVVLVLVIVGIGVLILLVRYRKCIKLKQVQSPPTETLGVENKGYFGDEQQIRTEPILPEIHFLNKPASKDSRGIADPNQSAK
Specifications

Specifications

AAI48736.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
109.89kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
ADAM7
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
8756
ADAM7 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EAPI/GP-83
ADAM7
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers