missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ADAMTSL1 Partial ORF (NP_644669.1, 192 a.a. - 291 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_644669.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 92949 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16181247
|
Abnova™
H00092949-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16171247
|
Abnova™
H00092949-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a secreted protein and member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motif) family. This protein lacks the metalloproteinase and disintegrin-like domains, which are typical of the ADAMTS family, but contains other ADAMTS domains, including the thrombospondin type 1 motif. This protein may have important functions in the extracellular matrix. Alternative splicing results in multiple transcript variants encoding distinct proteins. [provided by RefSeq]
Sequence: YKSQLSATKSDDTVVAIPYGSRHIRLVLKGPDHLYLETKTLQGTKGENSLNSTGTFLVDNSSVDFQKFPDKEILRMAGPLTADFIVKIRNSGSADSTVQFSpecifications
NP_644669.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ADAMTSR1/C9orf94/DKFZp686L03130/FLJ35283/FLJ41032/FLJ46891/MGC118803/MGC118805/MGC40193/PUNCTIN | |
ADAMTSL1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
92949 | |
ADAMTSL1 (Human) Recombinant Protein (Q01) | |
YKSQLSATKSDDTVVAIPYGSRHIRLVLKGPDHLYLETKTLQGTKGENSLNSTGTFLVDNSSVDFQKFPDKEILRMAGPLTADFIVKIRNSGSADSTVQF | |
RUO | |
ADAMTSL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |