Learn More
Abnova™ Human ADHFE1 Full-length ORF (NP_653251.1, 1 a.a. - 419 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00137872-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The ADHFE1 gene encodes hydroxyacid-oxoacid transhydrogenase (EC 1.1.99.24), which is responsible for the oxidation of 4-hydroxybutyrate in mammalian tissues (Kardon et al., 2006 [PubMed 16616524]).[supplied by OMIM]
Sequence: MAVSNIRYGAAVTKEVGMDLKNMGAKNVCLMTDKNLSKLPPVQVAMDSLVKNGIPFTVYDNVRVEPTDSSFMEAIEFAQKGAFDAYVAVGGGSTMDTCKAANLYASSPHSDFLDYVSAPIGKGKPVSVPLKPLIAVPTTSGTGSETTGVAIFDYEHLKVKIGITSRAIKPTLGLIDPLHTLHMPARVVANSGFDVLCHALESYTTLPYHLRSPCPSNPITRPAYQGSNPISDIWAIHALRIVAKYLKRAVRNPDDLEARSHMHLASAFAGIGFGNAGVHLCHGMSYPISGLVKMYKAKDYNVDHPLVPHGLSVVLTSPAVFTFTAQMFPERHLEMAEILGADTRTARIQDAGLVLADTLRKFLFDLDVDDGLAAVGYSKADIPALVKGTLPQERVTKLAPRPQSEEDLAALFEASMKLYSpecifications
NP_653251.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
71.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ADHFE1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
137872 | |
ADHFE1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ADH8/FLJ32430/HMFT2263/HOT/MGC48605 | |
ADHFE1 | |
Recombinant | |
wheat germ expression system |