missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human Adiponectin/Acrp30 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB10651-SP
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Adiponectin/Acrp30 Monoclonal specifically detects Adiponectin/Acrp30 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifikationer
| Adiponectin/Acrp30 | |
| Monoclonal | |
| LYOPH | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q15848 | |
| ADIPOQ | |
| Mouse myeloma cell line NS0-derived recombinant human Adiponectin Glu19-Asn244 Accession # Q15848 | |
| 25 μg | |
| Primary | |
| Detects an epitope in the globular domain (aa 104 - 244) of human Adiponectin. In ELISAs, this antibody shows no cross-reactivity or interference with recombinant mouse (rm) Adiponectin or rh6Ckine. | |
| Human | |
| Purified |
| ELISA | |
| 166126 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ACDC, Acrp30, ACRP30ADPN, adipocyte, C1Q and collagen domain containing, adiponectin, adiponectin, C1Q and collagen domain containing, AdipoQ, ADIPQTL1, apM-1, ApM1, APM1APM-1, C1q and collagen domain-containing protein, GBP28, GBP28apM1, Gelatin-binding protein | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9370 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering