Learn More
Abnova™ Human ADRA1A Partial ORF (NP_150646, 1 a.a. - 27 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000148-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Alpha-1-adrenergic receptors (alpha-1-ARs) are members of the G protein-coupled receptor superfamily. They activate mitogenic responses and regulate growth and proliferation of many cells. There are 3 alpha-1-AR subtypes: alpha-1A, -1B and -1D, all of which signal through the Gq/11 family of G-proteins and different subtypes show different patterns of activation. This gene encodes alpha-1A-adrenergic receptor. Alternative splicing of this gene generates four transcript variants, which encode four different isoforms with distinct C-termini but having similar ligand binding properties. [provided by RefSeq]
Sequence: MVFLSGNASDSSNCTQPPAPVNISKAISpecifications
NP_150646 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
28.71kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVFLSGNASDSSNCTQPPAPVNISKAI | |
RUO | |
ADRA1A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
148 | |
ADRA1A (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ADRA1C/ADRA1L1/ALPHA1AAR | |
ADRA1A | |
Recombinant | |
wheat germ expression system |