missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ADSSL1 Full-length ORF (NP_689541.1, 1 a.a. - 457 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_689541.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 122622 |
Molecular Weight (g/mol) | 76.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16147523
|
Abnova™
H00122622-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16137523
|
Abnova™
H00122622-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ADSSL1 is a muscle isozyme of adenylosuccinate synthase (EC 6.3.4.4), which catalyzes the initial reaction in the conversion of inosine monophosphate (IMP) to adenosine monophosphate (AMP) (Sun et al., 2005 [PubMed 15786719]).[supplied by OMIM]
Sequence: MSGTRASNDRPPGAGGVKRGRLQQEAAATGSRVTVVLGAQWGDEGKGKVVDLLATDADIISRCQGGNNAGHTVVVDGKEYDFHLLPSGIINTKAVSFIGNGVVIHLPGLFEEAEKNEKKGLKDWEKRLIISDRAHLVFDFHQAVDGLQEVQRQAQEGKNIGTTKKGIGPTYSSKAARTGLRICDLLSDFDEFSSRFKNLAHQHQSMFPTLEIDIEGQLKRLKGFAERIRPMVRDGVYFMYEALHGPPKKILVEGANAALLDIDFGTYPFVTSSNCTVGGVCTGLGIPPQNIGDVYGVVKAYTTRVGIGAFPTEQINEIGGLLQTRGHEWGVTTGRKRRCGWLDLMILRYAHMVNGFTALALTKLDILDVLGEVKVGVSYKLNGKRIPYFPANQEMLQKVEVEYETLPGWKADTTGARRWEDLPPQAQNYIRFVENHVGVAVKWVGVGKSRESMIQLFSpecifications
NP_689541.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
76.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ38602 | |
ADSSL1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
122622 | |
ADSSL1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
ADSSL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |