missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AGPAT3 Full-length ORF (AAH04219.1, 1 a.a. - 63 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00056894-P02.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16122303

  • 6845.00 SEK / 25µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is an acyltransferase that converts lysophosphatidic acid into phosphatidic acid, which is the second step in the de novo phospholipid biosynthetic pathway. The encoded protein may be an integral membrane protein. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]

Sequence: MGSPAHLPKDAIAVFIVFPRVRHWLSGTRRQCTRRAPYVRCFKVPVLRKRSLVEEMSAEQITK
Specifications

Specifications

AAH04219.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
AGPAT3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
56894
AGPAT3 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LPAAT-GAMMA1/MGC4604
AGPAT3
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers