Learn More
Abnova™ Human AIP (AAI04828, 1 a.a. - 330 a.a.) Full-length Recombinant Protein with His tag
Used for SDS-PAGE
Brand: Abnova™ P3724.100ug
Additional Details : Weight : 0.02000kg
Description
The protein encoded by this gene is a receptor for aryl hydrocarbons and a ligand-activated transcription factor. The encoded protein is found in the cytoplasm as part of a multiprotein complex, but upon binding of ligand is transported to the nucleus. This protein can regulate the expression of many xenobiotic metabolizing enzymes. Also, the encoded protein can bind specifically to and inhibit the activity of hepatitis B virus. [provided by RefSeq]
Sequence: MGSSHHHHHHSSGLVPRGSHMADIIARLREDGIQKRVIQEGRGELPDFQDGTKATFHYRTLHSDDEGTVLDDSRARGKPMELIIGKKFKLPVWETIVCTMREGEIAQFLCDIKHVVLYPLVAKSLRNIAVGKDPLEGQRHCCGVAQMREHSSLGHADLDALQQNPQPLIFHMEMLKVESPGTYQQDPWAMTDEEKAKAVPLIHQEGNRLYREGHVKEAAAKYYDAIACLKNLQMKEQPGSPEWIQLDKQITPLLLNYCQCKLVVEEYYEVLDHCSSILNKYDDNVKAYFKRGKAHAAVWNAQEAQADFAKVLELDPALAPVVSRELRALEARIRQKDEEDKARFRGIFSHSpecifications
AAI04828 | |
SDS-PAGE | |
9049 | |
AIP (Human) Recombinant Protein | |
Conventional Chromatography | |
Store at 4°C for 1-2 weeks. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARA9/FKBP16/FKBP37/SMTPHN/XAP2 | |
AIP | |
Recombinant | |
Escherichia coli expression system | |
>90% by SDS-PAGE |
1 mg/mL | |
In 20mM Tris-HCl, pH 8.0 (1mM dithiothreitol, 10% glycerol) | |
39.8kDa | |
Escherichia coli expression system | |
100 ug | |
RUO | |
AIP | |
E. coli | |
His | |
Liquid |