Learn More
Abnova™ Human ALAS2 Partial ORF (NP_000023, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000212-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The product of this gene specifies an erythroid-specific mitochondrially located enzyme. The encoded protein catalyzes the first step in the heme biosynthetic pathway. Defects in this gene cause X-linked pyridoxine-responsive sideroblastic anemia. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: MVTAAMLLQCCPVLARGPTSLLGKVVKTHQFLFGIGRCPILATQGPNCSQIHLKATKAGGDSPSWAKGHCPFMLSELQDGKSKIVQKAAPEVQEDVKAFKSpecifications
NP_000023 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVTAAMLLQCCPVLARGPTSLLGKVVKTHQFLFGIGRCPILATQGPNCSQIHLKATKAGGDSPSWAKGHCPFMLSELQDGKSKIVQKAAPEVQEDVKAFK | |
RUO | |
ALAS2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
212 | |
ALAS2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ALAS-E/ALASE/ANH1/ASB/FLJ93603/XLSA | |
ALAS2 | |
Recombinant | |
wheat germ expression system |