Learn More
Abnova™ Human ALDH1L1 Partial ORF (NP_036322, 803 a.a. - 902 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010840-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene catalyzes the conversion of 10-formyltetrahydrofolate, NADP, and water to tetrahydrofolate, NADPH, and carbon dioxide. The encoded protein belongs to the aldehyde dehydrogenase family and is responsible for formate oxidation in vivo. Deficiencies in this gene can result in an accumulation of formate and subsequent methanol poisoning. [provided by RefSeq]
Sequence: EESFGPVMIISRFADGDLDAVLSRANATEFGLASGVFTRDINKALYVSDKLQAGTVFVNTYNKTDVAAPFGGFKQSGFGKDLGEAALNEYLRVKTVTFEYSpecifications
NP_036322 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EESFGPVMIISRFADGDLDAVLSRANATEFGLASGVFTRDINKALYVSDKLQAGTVFVNTYNKTDVAAPFGGFKQSGFGKDLGEAALNEYLRVKTVTFEY | |
RUO | |
ALDH1L1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10840 | |
ALDH1L1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp781N0997/FTHFD | |
ALDH1L1 | |
Recombinant | |
wheat germ expression system |